2-(Isopropylamino)isonicotinic acid

Supplier: Matrix Scientific

052842-500MG 052842-2.5G
101901-256EA 364.62 USD
101901-256 101901-258
2-(Isopropylamino)isonicotinic acid
2-(Isopropylamino)isonicotinic acid

Hazard: Irritant

Formula: C₉H₁₂N₂O₂
MDL Number: MFCD11131982
CAS Number: 77314-51-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR