3-Chloro-N-isopropyl-5-(trifluoromethyl)-2-pyridinamine

Supplier: Matrix Scientific

052840-500MG 052840-2.5G
101901-248EA 364.62 USD
101901-248 101901-250
3-Chloro-N-isopropyl-5-(trifluoromethyl)-2-pyridinamine
3-Chloro-N-isopropyl-5-(trifluoromethyl)-2-pyridinamine

Hazard: Irritant

Formula: C₉H₁₀ClF₃N₂
MDL Number: MFCD11124523
CAS Number: 1036436-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR