6-Chloro-N-isopropyl-4-pyrimidinamine

Supplier: Matrix Scientific

Synonyms: 6-Chloro-N-isopropylpyrimidin-4-amine, 4-Chloro-6-(isopropylamino)pyrimidine

052839-500MG 052839-2.5G
101901-244EA 364.62 USD
101901-244 101901-246
6-Chloro-N-isopropyl-4-pyrimidinamine
6-Chloro-N-isopropyl-4-pyrimidinamine

Hazard: Irritant

Formula: C₇H₁₀ClN₃
MW: 171.63 g/mol
MDL Number: MFCD09756956
CAS Number: 945896-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR