5-Bromo-N-isopropyl-4-methyl-2-pyridinamine

Supplier: Matrix Scientific

052838-500MG 052838-2.5G
101901-240EA 364.62 USD
101901-240 101901-242
5-Bromo-N-isopropyl-4-methyl-2-pyridinamine
5-Bromo-N-isopropyl-4-methyl-2-pyridinamine

Hazard: Irritant

Formula: C₉H₁₃BrN₂
MDL Number: MFCD13562405
CAS Number: 1219957-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR