4-(Isopropylamino)-3-nitrobenzoic acid

Supplier: Matrix Scientific

052837-500MG 052837-2.5G
101901-236EA 364.62 USD
101901-236 101901-238
4-(Isopropylamino)-3-nitrobenzoic acid
4-(Isopropylamino)-3-nitrobenzoic acid

Hazard: Irritant

Formula: C₁₀H₁₂N₂O₄
MDL Number: MFCD08704414
CAS Number: 284672-95-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR