Ethyl-3-amino-4-(isopropylamino)benzoate

Supplier: Matrix Scientific

052836-500MG 052836-2.5G
101901-232EA 364.62 USD
101901-232 101901-234
Ethyl-3-amino-4-(isopropylamino)benzoate
Ethyl-3-amino-4-(isopropylamino)benzoate

Hazard: Irritant

Formula: C₁₂H₁₈N₂O₂
MDL Number: MFCD13562404
CAS Number: 1187570-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR