N-Allyl-4-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052833-500MG 052833-2.5G
101901-220EA 364.62 USD
101901-220 101901-222
N-Allyl-4-piperidinecarboxamide hydrochloride
N-Allyl-4-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₉H₁₇ClN₂O
MDL Number: MFCD13562402
CAS Number: 1219957-31-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR