N-Allyl-3-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052832-500MG 052832-2.5G
101901-216EA 364.62 USD
101901-216 101901-218
N-Allyl-3-piperidinecarboxamide hydrochloride
N-Allyl-3-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₉H₁₇ClN₂O
MDL Number: MFCD13562401
CAS Number: 1220027-49-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR