N-Allyl-2-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052831-500MG 052831-2.5G
101901-212EA 364.62 USD
101901-212 101901-214
N-Allyl-2-piperidinecarboxamide hydrochloride
N-Allyl-2-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₉H₁₇ClN₂O
MDL Number: MFCD13562400
CAS Number: 1236255-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR