N-Allyl-2-pyrrolidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052830-500MG 052830-2.5G
101901-208EA 364.62 USD
101901-208 101901-210
N-Allyl-2-pyrrolidinecarboxamide hydrochloride
N-Allyl-2-pyrrolidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₈H₁₅ClN₂O
MW: 190.67 g/mol
MDL Number: MFCD13562399
CAS Number: 1236254-98-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR