N-Allyl-2-amino-3-phenylpropanamide hydrochloride

Supplier: Matrix Scientific

052829-500MG 052829-2.5G
101901-204EA 364.62 USD
101901-204 101901-206
N-Allyl-2-amino-3-phenylpropanamide hydrochloride
N-Allyl-2-amino-3-phenylpropanamide hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂O
MDL Number: MFCD13562398
CAS Number: 1236261-01-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR