N-Allyl-2-(methylamino)acetamide hydrochloride

Supplier: Matrix Scientific

052825-500MG 052825-2.5G
101901-188EA 364.62 USD
101901-188 101901-190
N-Allyl-2-(methylamino)acetamide hydrochloride
N-Allyl-2-(methylamino)acetamide hydrochloride

Hazard: Irritant

Formula: C₆H₁₃ClN₂O
MDL Number: MFCD13562394
CAS Number: 1220034-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR