N-Allyl-2-aminopropanamide hydrochloride

Supplier: Matrix Scientific

052824-500MG 052824-2.5G
101901-184EA 364.62 USD
101901-184 101901-186
N-Allyl-2-aminopropanamide hydrochloride
N-Allyl-2-aminopropanamide hydrochloride

Hazard: Irritant

Formula: C₆H₁₃ClN₂O
MDL Number: MFCD13562393
CAS Number: 1236262-16-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR