N-Allyl-6-chloro-2-pyridinamine

Supplier: Matrix Scientific

052822-500MG 052822-2.5G
101901-176EA 364.62 USD
101901-176 101901-178
N-Allyl-6-chloro-2-pyridinamine
N-Allyl-6-chloro-2-pyridinamine

Hazard: Irritant

Formula: C₈H₉ClN₂
MDL Number: MFCD13562391
CAS Number: 791095-96-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR