N-Allyl-5-bromo-3-methyl-2-pyridinamine

Supplier: Matrix Scientific

052820-500MG 052820-2.5G
101901-168EA 364.62 USD
101901-168 101901-170
N-Allyl-5-bromo-3-methyl-2-pyridinamine
N-Allyl-5-bromo-3-methyl-2-pyridinamine

Hazard: Irritant

Formula: C₉H₁₁BrN₂
MDL Number: MFCD13562389
CAS Number: 1220018-63-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR