N2-Allyl-2,3-pyridinediamine

Supplier: Matrix Scientific

052818-500MG 052818-2.5G
101901-160EA 364.62 USD
101901-160 101901-162
N2-Allyl-2,3-pyridinediamine
N2-Allyl-2,3-pyridinediamine

Hazard: Irritant

Formula: C₈H₁₁N₃
MDL Number: MFCD11561637
CAS Number: 1082155-17-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR