2-(Allylamino)nicotinic acid

Supplier: Matrix Scientific

052817-500MG 052817-2.5G
101901-156EA 166.15 USD
101901-156 101901-158
2-(Allylamino)nicotinic acid
2-(Allylamino)nicotinic acid

Hazard: Irritant

Formula: C₉H₁₀N₂O₂
MDL Number: MFCD11131948
CAS Number: 1019126-98-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR