N-Allyl-5-bromo-4-methyl-2-pyridinamine

Supplier: Matrix Scientific

052813-500MG 052813-2.5G
101901-140EA 364.62 USD
101901-140 101901-142
N-Allyl-5-bromo-4-methyl-2-pyridinamine
N-Allyl-5-bromo-4-methyl-2-pyridinamine

Hazard: Irritant

Formula: C₉H₁₁BrN₂
MDL Number: MFCD13562386
CAS Number: 1220034-28-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR