N-Butyl-2-pyrrolidinecarboxamide hydrochloride
Supplier: Matrix Scientific
052805-500MG
052805-2.5G
101901-108EA
364.62
USD
101901-108
101901-110
N-Butyl-2-pyrrolidinecarboxamide hydrochloride
N-Butyl-2-pyrrolidinecarboxamide hydrochloride
Hazard: Irritant
Formula:
C₉H₁₉ClN₂O |
MDL Number:
MFCD13562381 CAS Number: 1236266-64-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...