2-Amino-N-butyl-3-phenylpropanamide hydrochloride

Supplier: Matrix Scientific

052804-500MG 052804-2.5G
101901-104EA 364.62 USD
101901-104 101901-106
2-Amino-N-butyl-3-phenylpropanamide hydrochloride
2-Amino-N-butyl-3-phenylpropanamide hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₁ClN₂O
MDL Number: MFCD13562380
CAS Number: 1236254-97-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR