3-Amino-N-butylpropanamide hydrochloride

Supplier: Matrix Scientific

052801-500MG 052801-2.5G
101901-092EA 364.62 USD
101901-092 101901-094
3-Amino-N-butylpropanamide hydrochloride
3-Amino-N-butylpropanamide hydrochloride

Hazard: Irritant

Formula: C₇H₁₇ClN₂O
MDL Number: MFCD13562377
CAS Number: 1220034-22-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR