N-Butyl-2-(methylamino)acetamide hydrochloride
Supplier: Matrix Scientific
052800-500MG
052800-2.5G
101901-088EA
364.62
USD
101901-088
101901-090
N-Butyl-2-(methylamino)acetamide hydrochloride
N-Butyl-2-(methylamino)acetamide hydrochloride
Hazard: Irritant
Formula:
C₇H₁₇ClN₂O |
MDL Number:
MFCD13562376 CAS Number: 262300-46-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...