N-Butyl-6-chloro-2-pyridinamine

Supplier: Matrix Scientific

052797-500MG 052797-2.5G
101901-076EA 364.62 USD
101901-076 101901-078
N-Butyl-6-chloro-2-pyridinamine
N-Butyl-6-chloro-2-pyridinamine

Hazard: Irritant

Formula: C₉H₁₃ClN₂
MDL Number: MFCD13562373
CAS Number: 3298-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR