N-Butyl-2-chloronicotinamide

Supplier: Matrix Scientific

052794-500MG 052794-2.5G
101901-064EA 364.62 USD
101901-064 101901-066
N-Butyl-2-chloronicotinamide
N-Butyl-2-chloronicotinamide

Hazard: Irritant

Formula: C₁₀H₁₃ClN₂O
MDL Number: MFCD01758482
CAS Number: 56149-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR