N2-Butyl-2,3-pyridinediamine

Supplier: Matrix Scientific

052793-500MG 052793-2.5G
101901-060EA 364.62 USD
101901-060 101901-062
N2-Butyl-2,3-pyridinediamine
N2-Butyl-2,3-pyridinediamine

Hazard: Irritant

Formula: C₉H₁₅N₃
MDL Number: MFCD11101082
CAS Number: 53929-75-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR