2-(Butylamino)isonicotinic acid

Supplier: Matrix Scientific

052791-500MG 052791-2.5G
101901-052EA 364.62 USD
101901-052 101901-054
2-(Butylamino)isonicotinic acid
2-(Butylamino)isonicotinic acid

Hazard: Irritant

Formula: C₁₀H₁₄N₂O₂
MDL Number: MFCD11132079
CAS Number: 77314-78-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR