N-Butyl-3-chloro-5-(trifluoromethyl)-2-pyridinamine

Supplier: Matrix Scientific

Synonyms: 2-(Butylamino)-3-chloro-5-(trifluoromethyl)pyridine

052789-500MG 052789-2.5G
101901-044EA 364.62 USD
101901-044 101901-046
N-Butyl-3-chloro-5-(trifluoromethyl)-2-pyridinamine
N-Butyl-3-chloro-5-(trifluoromethyl)-2-pyridinamine

Hazard: Irritant

Formula: C₁₀H₁₂ClF₃N₂
MW: 252.67 g/mol
MDL Number: MFCD11119963
CAS Number: 1041535-26-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR