N-Butyl-6-chloro-4-pyrimidinamine

Supplier: Matrix Scientific

Synonyms: 4-(Butylamino)-6-chloropyrimidine

052788-500MG 052788-2.5G
101901-040EA 364.62 USD
101901-040 101901-042
N-Butyl-6-chloro-4-pyrimidinamine
N-Butyl-6-chloro-4-pyrimidinamine

Hazard: Irritant

Formula: C₈H₁₂ClN₃
MW: 185.66 g/mol
MDL Number: MFCD09800955
CAS Number: 26423-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR