4-(Butylamino)-3-nitrobenzoic acid
Supplier: Matrix Scientific
052786-500MG
052786-2.5G
101901-032EA
364.62
USD
101901-032
101901-034
4-(Butylamino)-3-nitrobenzoic acid
4-(Butylamino)-3-nitrobenzoic acid
Hazard: Irritant
Formula:
C₁₁H₁₄N₂O₄ |
MDL Number:
MFCD00434901 CAS Number: 120321-65-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...