Ethyl-3-amino-4-(butylamino)benzoate

Supplier: Matrix Scientific

052785-500MG 052785-2.5G
101901-028EA 364.62 USD
101901-028 101901-030
Ethyl-3-amino-4-(butylamino)benzoate
Ethyl-3-amino-4-(butylamino)benzoate

Hazard: Irritant

Formula: C₁₃H₂₀N₂O₂
MDL Number: MFCD13344028
CAS Number: 1220018-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR