N-Propyl-4-piperidinecarboxamide hydrochloride
Supplier: Matrix Scientific
052782-500MG
052782-2.5G
101901-016EA
364.62
USD
101901-016
101901-018
N-Propyl-4-piperidinecarboxamide hydrochloride
N-Propyl-4-piperidinecarboxamide hydrochloride
Hazard: Irritant
Formula:
C₉H₁₉ClN₂O |
MDL Number:
MFCD13562369 CAS Number: 1220036-87-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...