N-Propyl-3-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052781-500MG 052781-2.5G
101901-012EA 364.62 USD
101901-012 101901-014
N-Propyl-3-piperidinecarboxamide hydrochloride
N-Propyl-3-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₉H₁₉ClN₂O
MDL Number: MFCD13562368
CAS Number: 1220034-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR