N-Propyl-2-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052780-500MG 052780-2.5G
101901-008EA 364.62 USD
101901-008 101901-010
N-Propyl-2-piperidinecarboxamide hydrochloride
N-Propyl-2-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₉H₁₉ClN₂O
MDL Number: MFCD13562367
CAS Number: 444992-97-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR