2-Amino-3-methyl-N-propylbutanamide hydrochloride

Supplier: Matrix Scientific

052776-500MG 052776-2.5G
101900-992EA 364.62 USD
101900-992 101900-994
2-Amino-3-methyl-N-propylbutanamide hydrochloride
2-Amino-3-methyl-N-propylbutanamide hydrochloride

Hazard: Irritant

Formula: C₈H₁₉ClN₂O
MW: 194.71 g/mol
MDL Number: MFCD13562363
CAS Number: 1236263-44-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR