2-(Methylamino)-N-propylacetamide hydrochloride

Supplier: Matrix Scientific

052774-500MG 052774-2.5G
101900-984EA 364.62 USD
101900-984 101900-986
2-(Methylamino)-N-propylacetamide hydrochloride
2-(Methylamino)-N-propylacetamide hydrochloride

Hazard: Irritant

Formula: C₆H₁₅ClN₂O
MW: 166.65 g/mol
MDL Number: MFCD07287585
CAS Number: 1049764-41-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR