6-Chloro-N-propyl-2-pyridinamine
Supplier: Matrix Scientific
052771-500MG
052771-2.5G
101900-972EA
364.62
USD
101900-972
101900-974
6-Chloro-N-propyl-2-pyridinamine
6-Chloro-N-propyl-2-pyridinamine
Hazard: Irritant
Formula:
C₈H₁₁ClN₂ |
MDL Number:
MFCD13562359 CAS Number: 40263-65-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...