N-(5-Bromo-3-methyl-2-pyridinyl)-N-propylamine

Supplier: Matrix Scientific

052769-500MG 052769-2.5G
101900-964EA 364.62 USD
101900-964 101900-966
N-(5-Bromo-3-methyl-2-pyridinyl)-N-propylamine
N-(5-Bromo-3-methyl-2-pyridinyl)-N-propylamine

Hazard: Irritant

Formula: C₉H₁₃BrN₂
MDL Number: MFCD13562358
CAS Number: 1219982-70-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR