N2-Propyl-2,3-pyridinediamine
Supplier: Matrix Scientific
052767-500MG
052767-2.5G
101900-956EA
364.62
USD
101900-956
101900-958
N2-Propyl-2,3-pyridinediamine
N2-Propyl-2,3-pyridinediamine
Hazard: Irritant
Formula:
C₈H₁₃N₃ |
MDL Number:
MFCD08692148 CAS Number: 439571-80-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...