2-(Propylamino)nicotinic acid

Supplier: Matrix Scientific

052766-500MG 052766-2.5G
101900-952EA 364.62 USD
101900-952 101900-954
2-(Propylamino)nicotinic acid
2-(Propylamino)nicotinic acid

Hazard: Irritant

Formula: C₉H₁₂N₂O₂
MDL Number: MFCD11131961
CAS Number: 74611-52-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR