2-(Propylamino)isonicotinic acid

Supplier: Matrix Scientific

052765-500MG 052765-2.5G
101900-948EA 384.62 USD
101900-948 101900-950
2-(Propylamino)isonicotinic acid
2-(Propylamino)isonicotinic acid

Hazard: Irritant

Formula: C₉H₁₂N₂O₂
MW: 180.21 g/mol
MDL Number: MFCD11131957
CAS Number: 1019127-04-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR