3-Chloro-N-propyl-5-(trifluoromethyl)-2-pyridinamine

Supplier: Matrix Scientific

052763-500MG 052763-2.5G
101900-940EA 364.62 USD
101900-940 101900-942
3-Chloro-N-propyl-5-(trifluoromethyl)-2-pyridinamine
3-Chloro-N-propyl-5-(trifluoromethyl)-2-pyridinamine

Hazard: Irritant

Formula: C₉H₁₀ClF₃N₂
MW: 238.64 g/mol
MDL Number: MFCD11119991
CAS Number: 1041557-60-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR