6-Chloro-N-propyl-4-pyrimidinamine

Supplier: Matrix Scientific

Synonyms: 4-Chloro-6-(propylamino)pyrimidine

052762-500MG 052762-2.5G
101900-936EA 364.62 USD
101900-936 101900-938
6-Chloro-N-propyl-4-pyrimidinamine
6-Chloro-N-propyl-4-pyrimidinamine

Hazard: Irritant

Formula: C₇H₁₀ClN₃
MW: 171.63 g/mol
MDL Number: MFCD09475893
CAS Number: 941294-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR