N-(5-Bromo-4-methyl-2-pyridinyl)-N-propylamine

Supplier: Matrix Scientific

052761-500MG 052761-2.5G
101900-932EA 364.62 USD
101900-932 101900-934
N-(5-Bromo-4-methyl-2-pyridinyl)-N-propylamine
N-(5-Bromo-4-methyl-2-pyridinyl)-N-propylamine

Hazard: Irritant

Formula: C₉H₁₃BrN₂
MDL Number: MFCD13562357
CAS Number: 1220034-12-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR