3-Nitro-4-(propylamino)benzoic acid

Supplier: Matrix Scientific

052760-500MG 052760-2.5G
101900-928EA 364.62 USD
101900-928 101900-930
3-Nitro-4-(propylamino)benzoic acid
3-Nitro-4-(propylamino)benzoic acid

Hazard: Irritant

Formula: C₁₀H₁₂N₂O₄
MDL Number: MFCD02089273
CAS Number: 68740-31-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR