Ethyl-3-amino-4-(propylamino)benzoate
Supplier: Matrix Scientific
052759-500MG
052759-2.5G
101900-924EA
364.62
USD
101900-924
101900-926
Ethyl-3-amino-4-(propylamino)benzoate
Ethyl-3-amino-4-(propylamino)benzoate
Hazard: Irritant
Formula:
C₁₂H₁₈N₂O₂ |
MDL Number:
MFCD13562356 CAS Number: 848819-85-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...