4-Piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

Synonyms: Piperidine-4-carboxamide hydrochloride

052757-500MG 052757-2.5G
101900-916EA 364.62 USD
101900-916 101900-918
4-Piperidinecarboxamide hydrochloride
4-Piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₆H₁₃ClN₂O
MW: 164.64 g/mol
MDL Number: MFCD11615743
CAS Number: 39674-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR