3-Piperidinecarboxamide hydrochloride
Supplier: Matrix Scientific
052756-500MG
052756-2.5G
101900-912EA
364.62
USD
101900-912
101900-914
3-Piperidinecarboxamide hydrochloride
3-Piperidinecarboxamide hydrochloride
Hazard: Irritant
Formula:
C₆H₁₃ClN₂O |
MDL Number:
MFCD09037931 CAS Number: 828300-58-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...