DL-Phenylalanine amide hydrochloride
Supplier: Matrix Scientific
Synonyms:
H-DL-Phe-NH₂·HCl, H-DL-Phe-NH2.HCl
052753-500MG
052753-2.5G
101900-900EA
364.62
USD
101900-900
101900-902
DL-Phenylalanine amide hydrochloride
DL-Phenylalanine amide hydrochloride
Hazard: Irritant
Formula:
C₉H₁₃ClN₂O MW: 200.666 g/mol |
MDL Number:
MFCD00070243 CAS Number: 108321-83-1 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...