2-Amino-2-methylpropanamide hydrochloride

Supplier: Matrix Scientific

052752-500MG 052752-2.5G
101900-896EA 384.62 USD
101900-896 101900-898
2-Amino-2-methylpropanamide hydrochloride
2-Amino-2-methylpropanamide hydrochloride

Hazard: Irritant

Formula: C₄H₁₁ClN₂O
MDL Number: MFCD12913069
CAS Number: 17704-74-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR