2-(methylamino)acetamide hydrochloride

Supplier: Matrix Scientific

Synonyms: H-Sar-NH₂·HCl, H-Sar-NH2.HCl

052749-500MG 052749-2.5G
101900-884EA 364.62 USD
101900-884 101900-886
2-(methylamino)acetamide hydrochloride
2-(methylamino)acetamide hydrochloride

Hazard: Irritant

Formula: C₃H₈N₂O·HCl
MW: 124.568 g/mol
MDL Number: MFCD00058283
CAS Number: 5325-64-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR